missing translation for 'onlineSavingsMsg'
Learn More
Tocris Bioscience™ INCB 024360-analog, >99% Product Code.: 30000881

Tocris Bioscience™ INCB 024360-analog, >99%
GREENER_CHOICE

Product Code. 30000881
10 mg, 10mg
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30000881

Brand: Tocris Bioscience™ 6007/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent indoleamine 2,3-dioxygenase (IDO) inhibitor

Potent indoleamine 2,3-dioxygenase (IDO) inhibitor (IC50 values are 19 and 67 nM in enzymatic activity and HeLa cell assays, respectively). Decreases kynurenine levels in plasma and reduces tumor growth in vivo. Cell membrane permeable.
Greener Choice Claims
  • Environmental benefits include:
    • Ozone-Safe or Safe for the Ozone Layer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 914471-09-3
Molecular Formula C9H7ClFN5O2

Specifications

Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 100mM in ethanol
Formula Weight 271.64
Purity >99%
Molecular Weight (g/mol) 271.636
Chemical Name or Material 4-Amino-N'-(3-chloro-4-fluorophenyl)-N-hydroxy-1,2,5-oxadiazole-3-carboximidamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tocris Bioscience™ INCB 024360-analog, >99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.