accessibility menu, dialog, popup

UserName

CAS RN 914471-09-3

HNNOHNNH2NOClF(E/Z)

CAS RN 914471-09-3

IUPAC Name: 4-{[(3-chloro-4-fluorophenyl)amino](nitroso)methylidene}-4,5-dihydro-1,2,5-oxadiazol-3-amine
Synonyms: 4-{[(3-chloro-4-fluorophenyl)amino](nitroso)methylidene}-5H-1,2,5-oxadiazol-3-amine
Molecular Weight (g/mol): 271.64
Molecular Formula: C9H7ClFN5O2
InChi Key: MUVQOOPKPBEAJZ-UHFFFAOYSA-N
SMILES: NC1=NONC1=C(NC1=CC(Cl)=C(F)C=C1)N=O
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results