CAS RN 172889-26-8
CAS RN 172889-26-8
IUPAC Name:
1-tert-butyl-3-(4-methylphenyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine
Synonyms:
1-tert-butyl-3-(4-methylphenyl)pyrazolo[3,4-d]pyrimidin-4-amine
Molecular Weight (g/mol):
281.36
Molecular Formula:
C16H19N5
InChi Key:
ZVPDNRVYHLRXLX-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)C1=NN(C2=NC=NC(N)=C12)C(C)(C)C
1
–
3
of
3
results
2
PP1, MedChemExpress
MedChemExpress PP1 is a potent, and Src family-selective tyrosine kinase inhibitor with IC50 of 5 and 6 nM for Lck and Fyn, respectively.
3