Learn More
Tocris Bioscience™ PP 1
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Potent inhibitor of Src-family tyrosine kinases. Inhibits p56lck and p59fynT (IC50 values are 5 and 6 nM respectively). Displays > 8000-fold selectivity over ZAP-70 and JAK2. Also moderately inhibits p38, CSK, PDGF receptors, RET-derived oncoproteins, c-Kit and Bcr-Abl.
Specifications
Specifications
| CAS | 172889-26-8 |
| Quantity | 10 mg |
| Molecular Formula | C16H19N5 |
| Formula Weight | 281.36 |
| Purity | >99% |
| Molecular Weight (g/mol) | 281.363 |
| Chemical Name or Material | PP 1 |