missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies PP1
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Description
- PP1 is a reversible inhibitor of the SRC family of tyrosine kinases
- It inhibits LCK, FYN, HCK and SRC with IC50 values of 5, 6, 20 and 170 nM, respectively (Hanke et al.) It is relatively selective for SRC family kinases versus other kinases, inhibiting epidermal growth factor receptor (EGFR), janus-activated kinase 2 (JAK2) and zeta-chain-associated protein kinase 70 (ZAP70) with IC50 values of 0.25, > 50, and > 100 μM, respectively, and KIT, platelet-derived growth factor receptor (PDGFR), and RET tyrosine kinase in the 75 - 100 nM range (Carlomagno et al.; Tatton et al.; Waltenberger et al.; Hanke et al.) PP1 also blocks TGF-β-mediated cellular responses by directly inhibiting type I TGF-β receptors (IC50 = 50 nM; Ungefroren et al.; Maeda et al.)
Specifications
Specifications
| CAS | 172889-26-8 |
| Synonym | AGL 1872; EI 275 |
| Quantity | 10 mg |
| Molecular Formula | C16H19N5 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction