Learn More
PP1, MedChemExpress
Product Code. 16452017
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16452017
Brand: Medchem Express HY13804
MedChemExpress PP1 is a potent, and Src family-selective tyrosine kinase inhibitor with IC50 of 5 and 6 nM for Lck and Fyn, respectively.
PP1 is a potent, and Src family-selective tyrosine kinase inhibitor with IC50 of 5 and 6 nM for Lck and Fyn, respectively.
Specifications
Chemical Name or Material PP1
Physical Form Solid
CAS 172889-26-8
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C16H19N5
For Use With (Application) Cancer-Kinase/protease
Synonym AGL 1872 EI 275
Solubility Information DMSO : 28 mg/mL (99.52 mM; Need ultrasonic)
SMILES NC1=C2C(N(C(C)(C)C)N=C2C3=CC=C(C)C=C3)=NC=N1
Molecular Weight (g/mol) 281.36
Formula Weight 281.36
Percent Purity 98.62%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.