accessibility menu, dialog, popup

UserName

CAS RN 97548-97-5

ClClCH3N(R)NNH2N(R)HH

CAS RN 97548-97-5

IUPAC Name: dihydrogen (5aR,9aR)-6-propyl-5H,5aH,6H,7H,8H,9H,9aH,10H-pyrido[2,3-g]quinazolin-2-amine dichloride
Synonyms: quinelorane
Molecular Weight (g/mol): 319.27
Molecular Formula: C14H24Cl2N4
InChi Key: WDEMLQIGYYLRRX-OWVUFADGSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CCCN1CCC[C@@H]2CC3=NC(N)=NC=C3C[C@@H]12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results