missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Quinelorane hydrochloride

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Dopamine D2 and D3 receptor agonist; Ki values are 5.7 and 3.4 nM respectively.
Specifications
Specifications
| CAS | 97548-97-5 |
| Quantity | 10 mg |
| Molecular Formula | C14H24Cl2N4 |
| Formula Weight | 319.27 |
| Purity | >99% |
| Molecular Weight (g/mol) | 319.274 |
| Chemical Name or Material | Quinelorane hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction