accessibility menu, dialog, popup

UserName

CAS RN 922-32-7

NaNaCH3NHOONH2NPOOO(E/Z)

CAS RN 922-32-7

IUPAC Name: disodium ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate
Synonyms: disodium phosphocreatine
Molecular Weight (g/mol): 255.08
Molecular Formula: C4H8N3Na2O5P
InChi Key: RNTXMYSPASRLFT-UHFFFAOYSA-L
SMILES: [Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results