missing translation for 'onlineSavingsMsg'
Learn More

Creatine Phosphate, Disodium Salt Tetrahydrate, MP Biomedicals™

Product Code. 11497570 Shop All MP Biomedicals Products
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 11497570

Brand: MP Biomedicals 02100520.2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Creatine Phosphate, Disodium Salt Tetrahydrate, MP Biomedicals is a substrate for CPK determination.

TRUSTED_SUSTAINABILITY

Specifications

CAS 922-32-7
pH 7 to 9 (1% aq. soln.)
Molecular Formula C4H8N3Na2O5P
MDL Number MFCD00150192
Quantity 250 mg
Synonym creatine phosphate sodium salt
Solubility Information Soluble in water (150mg/mL, clear, colorless solution).
InChI Key RNTXMYSPASRLFT-UHFFFAOYSA-L
SMILES [Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O
IUPAC Name disodium ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate
Molecular Weight (g/mol) 255.08
PubChem CID 131887655
Formula Weight 327.2
Percent Purity ≥98%
Physical Form Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.