CAS RN 289499-45-2
CAS RN 289499-45-2
IUPAC Name:
N-{4-[(3-chloro-4-fluorophenyl)amino]-7-[3-(morpholin-4-yl)propoxy]quinazolin-6-yl}prop-2-enamide dihydrochloride
Synonyms:
canertinib dihydrochloride
Molecular Weight (g/mol):
558.86
Molecular Formula:
C24H27Cl3FN5O3
InChi Key:
JZZFDCXSFTVOJY-UHFFFAOYSA-N
SMILES:
Cl.Cl.FC1=CC=C(NC2=C3C=C(NC(=O)C=C)C(OCCCN4CCOCC4)=CC3=NC=N2)C=C1Cl
1
–
5
of
5
results
1
Canertinib dihydrochloride, Thermo Scientific™
CAS: 289499-45-2 Molecular Formula: C24 H25 Cl F N5 O3 · 2 H Cl
2
Tocris Bioscience™ Canertinib dihydrochloride
Potent EGFR kinase inhibitor
3
Canertinib dihydrochloride, MedChemExpress
MedChemExpress Canertinib dihydrochloride (CI-1033 dihydrochloride) is a potent and irreversible EGFR inhibitor; inhibits cellular EGFR and ErbB2 autophosphorylation with IC50s of 7.4 and 9 nM. Canertinib dihydrochloride is active against vaccinia virus respiratory infection in mice.
4
5