Learn More
Canertinib dihydrochloride, Thermo Scientific™
Product Code. 17555565 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 17555565
Brand: Thermo Scientific Acros 468932500
Request Bulk or Custom Format
Chemical Identifiers
CAS 289499-45-2
Molecular Formula C24 H25 Cl F N5 O3 · 2 H Cl
Specifications
Quantity 250 mg
Formula Weight 558.87
MDL Number MFCD09954112
Physical Form Powder
Color White to Beige
Purity ≥97.5%
InChI Key JZZFDCXSFTVOJY-UHFFFAOYSA-N
SMILES Cl.Cl.FC1=CC=C(NC2=C3C=C(NC(=O)C=C)C(OCCCN4CCOCC4)=CC3=NC=N2)C=C1Cl
IUPAC Name N-{4-[(3-chloro-4-fluorophenyl)amino]-7-[3-(morpholin-4-yl)propoxy]quinazolin-6-yl}prop-2-enamide dihydrochloride
Molecular Weight (g/mol) 558.86
Chemical Name or Material Canertinib dihydrochloride