missing translation for 'onlineSavingsMsg'
Learn More

Canertinib dihydrochloride, MedChemExpress

Product Code. p-200098667
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16494926

Brand: Medchem Express HY10367A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Canertinib dihydrochloride (CI-1033 dihydrochloride) is a potent and irreversible EGFR inhibitor; inhibits cellular EGFR and ErbB2 autophosphorylation with IC50s of 7.4 and 9 nM. Canertinib dihydrochloride is active against vaccinia virus respiratory infection in mice.

Canertinib dihydrochloride (CI-1033 dihydrochloride) is a potent and irreversible EGFR inhibitor

Specifications

Chemical Name or Material Canertinib dihydrochloride
CAS 289499-45-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H27Cl3FN5O3
Quantity 100 mg
Synonym CI-1033 dihydrochloride PD-183805 dihydrochloride
Solubility Information DMSO : 62.5 mg/mL (111.83 mM; Need ultrasonic) ∣H2O : 25 mg/mL (44.73 mM; Need ultrasonic)
SMILES C=CC(NC1=CC2=C(N=CN=C2C=C1OCCCN3CCOCC3)NC4=CC=C(C(Cl)=C4)F)=O.[H]Cl.[H]Cl
Molecular Weight (g/mol) 558.86
Formula Weight 558.86
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.