CAS RN 20380-11-4
CAS RN 20380-11-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
6
of
6
results
1
(25R)-26-Hydroxycholesterol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Tocris Bioscience™ 27-Hydroxycholesterol
LXR agonist; also ER partial agonist
3
(25R)-26-Hydroxycholesterol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
(25R)-26-Hydroxycholesterol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
5
27-Hydroxycholesterol, MedChemExpress
MedChemExpress 27-Hydroxycholesterol is a selective estrogen receptor modulator and an agonist of the liver X receptor.
6