Learn More
Tocris Bioscience™ 27-Hydroxycholesterol
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
27-Hydroxycholesterol is a liver X receptor (LXR) agonist. Also estrogen receptor partial agonist. Suppresses β-estradiol -mediated breast cancer cell proliferation in vitro. Also increases α-synuclein levels and induces apoptosis in human neuroblastoma cells in vitro. Endogenous cholesterol metabolite.
Specifications
Specifications
| Chemical Name or Material | (3β,25R)-Cholest-5-ene-3,26-diol |
| CAS | 20380-11-4 |
| Synonym | 27 OHC, 27 HC |
| Quantity | 10 mg |
| Target | LXR-like Receptor Agonists |
| Molecular Formula | C27H46O2 |
| Formula Weight | 402.65 |
| Purity | >98% |