Learn More
(25R)-26-Hydroxycholesterol, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | (25R)-26-Hydroxycholesterol |
| CAS | 20380-11-4 |
| Molecular Formula | C27 H46 O2 |
| Quantity | 10 mg |
| Synonym | Cholest-5-ene-3,26-diol, (3β,25R)- (9CI, ACI), (3β,25R)-Cholest-5-ene-3,26-diol (ACI), Cholest-5-ene-3β,26-diol, (25R)- (8CI), (25R)-26-Hydroxycholesterol, (25R)-Cholest-5-ene-3β,26-diol, (3β,25R)-Cholest-5-ene-3,26-diol, 27-Hydroxy-Cholesterol |
| InChI Formula | InChI=1S/C27H46O2/c1-18(17-28)6-5-7-19(2)23-10-11-24-22-9-8-20-16-21(29)12-14-26(20,3)25(22)13-15-27(23,24)4/h8,18-19,21-25,28-29H,5-7,9-17H2,1-4H3/t18-,19-,21+,22+,23-,24+,25+,26+,27-/m1/s1 |
| SMILES | C[C@@H](CO)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC Name | (3S,8S,9S,10R,13R,14S,17R)-17-[(1R,5R)-6-hydroxy-1,5-dimethyl-hexyl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol |
| Molecular Weight (g/mol) | 402.652 |
| Formula Weight | 402.3498 |