Learn More
DWK Life Sciences Wheaton™ 12 x 32 mm E-Z Vial Convenience Pack With 11 mm Crimp Finish And Clear Vial With Patch
Wheaton E-Z Vials have a 40% larger opening improves sample accessibility while reducing autosampler. Writing patch allows for identification needle damage.
575.00 NOK - 1035.00 NOK
Specifications
Color | Clear |
---|---|
Diameter (Metric) Outer | 12 mm |
Material | Borosilicate Glass |
Volume (Metric) | 1.8 mL |
For Use With (Application) | For chromatography |
Product Code | Brand | Closure Color | Septum | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Color | Septum | Price | Quantity & Availability | ||||
12434931
|
DWK Life Sciences
W225174-0207 |
Green | PTFE, Silicone |
900.00 NOK
Pack of 100 |
Estimated Shipment: 30-09-2024
Log in to see stock. |
||||
12464931
|
DWK Life Sciences
W225174-0304 |
Blue | PTFE, Silicone, PTFE |
1025.00 NOK
Pack of 100 |
Estimated Shipment: 30-09-2024
Log in to see stock. |
||||
12424931
|
DWK Life Sciences
W225174-0204 |
Blue | PTFE, Silicone |
900.00 NOK
Pack of 100 |
Estimated Shipment: 30-09-2024
Log in to see stock. |
||||
12454931
|
DWK Life Sciences
W225174-0303 |
Red | PTFE, Silicone, PTFE |
1035.00 NOK
Pack of 100 |
Estimated Shipment: 30-09-2024
Log in to see stock. |
||||
12414931
|
DWK Life Sciences
W225174-0203 |
Red | PTFE, Silicone |
895.00 NOK
Pack of 100 |
Estimated Shipment: 30-09-2024
Log in to see stock. |
||||
12404931
|
DWK Life Sciences
W225174-02 |
Natural | PTFE, Silicone |
900.00 NOK
Pack of 100 |
Estimated Shipment: 30-09-2024
Log in to see stock. |
||||
12494921
|
DWK Life Sciences
W225174-0107 |
Green | PTFE, Rubber |
575.00 NOK
Pack of 100 |
Estimated Shipment: 30-09-2024
Log in to see stock. |
||||
12474921
|
DWK Life Sciences
W225174-0103 |
Red | PTFE, Rubber |
575.00 NOK
Pack of 100 |
Estimated Shipment: 30-09-2024
Log in to see stock. |
||||
12484921
|
DWK Life Sciences
W225174-0104 |
Blue | PTFE, Rubber |
575.00 NOK
Pack of 100 |
Estimated Shipment: 30-09-2024
Log in to see stock. |
||||
12464921
|
DWK Life Sciences
W225174-01 |
Natural | PTFE, Rubber |
575.00 NOK
Pack of 100 |
Estimated Shipment: 30-09-2024
Log in to see stock. |
||||
Description
- Manufactured from certified clear Type I borosilicate glass
- Writing patch
- Vials are packaged 100 per shelf pack and 10 packs per case
- 40% larger opening improves sample accessibility while reducing autosampler needle damage
Specifications
Clear | |
Borosilicate Glass | |
For chromatography | |
Aluminum | |
32 mm | |
11 mm |
12 mm | |
1.8 mL | |
1.25 in. | |
Crimp Cap | |
100 Cs. | |
Yes |