Learn More
DWK Life Sciences Wheaton™ 12 x 32 mm E-Z Vial Convenience Pack With 11 mm Crimp Finish And Clear Vial With Patch
Shop All Wheaton Science Products Products
Click to view available options
Closure Color:
Blue
Green
Natural
Red
Septum:
PTFE, Rubber
PTFE, Silicone
PTFE, Silicone, PTFE
Unit Size:
Pack of 100
Description
- Manufactured from certified clear Type I borosilicate glass
- Writing patch
- Vials are packaged 100 per shelf pack and 10 packs per case
- 40% larger opening improves sample accessibility while reducing autosampler needle damage
Specifications
Specifications
| Closure Color | Green |
| Color | Clear |
| Diameter (Metric) Outer | 12 mm |
| Material | Borosilicate Glass |
| Volume (Metric) | 1.8 mL |
| Septum | PTFE, Silicone |
| For Use With (Application) | Chromatography |
| Height (English) | 1.25 in. |
| Closure Material | Aluminum |
| Closure Type | Crimp Cap |
| Show More |