Learn More
DWK Life Sciences Wheaton™ 12 x 32 mm E-Z Vial Convenience Pack With 11 mm Crimp Finish And Clear Vial With Patch
Wheaton E-Z Vials have a 40% larger opening improves sample accessibility while reducing autosampler. Writing patch allows for identification needle damage.
Brand:  DWK Life Sciences W225174-0107
Additional Details : Weight : 0.45360kg
 View more versions of this product
Product Code. 12494921
  • 575.00 NOK / Pack of 100
Estimated Shipment: 27-09-2024
Description
  • Manufactured from certified clear Type I borosilicate glass
  • Writing patch
  • Vials are packaged 100 per shelf pack and 10 packs per case
  • 40% larger opening improves sample accessibility while reducing autosampler needle damage
Specifications
Specifications
Green
12 mm
1.8 mL
For chromatography
Aluminum
32 mm
11 mm
Clear
Borosilicate Glass
PTFE, Rubber
1.25 in.
Crimp Cap
100 Cs.