Learn More
SP Bel-Art™ Stirring Bars Spinbar™
Multi-faceted shape provides greater surface area and added turbulence for more vigorous, thorough mixing when compared to the smooth surface of cylindrical bars
93.10 NOK - 403.00 NOK
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
16438485
|
SP Bel-Art™
F37110-1308 |
13 mm | 8 mm |
167.00 NOK
Pack of 10 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
16458485
|
SP Bel-Art™
F37110-2208 |
22 mm | 8 mm |
288.00 NOK
Pack of 10 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
16468485
|
SP Bel-Art™
F37110-2508 |
25 mm | 8 mm |
288.00 NOK
Pack of 10 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
16488485
|
SP Bel-Art™
F37110-3808 |
38 mm | 8 mm |
377.00 NOK
Pack of 10 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
16498485
|
SP Bel-Art™
F37110-3810 |
38 mm | 10 mm |
403.00 NOK
Pack of 10 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
16408495
|
SP Bel-Art™
F37110-5108 |
51 mm | 8 mm |
211.00 NOK
Pack of 5 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
16418495
|
SP Bel-Art™
F37110-5110 |
51 mm | 10 mm |
339.00 NOK
Pack of 5 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
16448485
|
SP Bel-Art™
F37110-1608 |
16 mm | 8 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
16478485
|
SP Bel-Art™
F37110-2908 |
29 mm | 8 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
16428495
|
SP Bel-Art™
F37110-7613 |
76 mm | 13 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- With pivot ring for better mixing conditions
- Alnico magnet encapsulated with PTFE™ brand PTFE coating
- FDA Grade, USP Class VI
- Good chemical compatibility
- Autoclavable
Specifications
Octagon |