Learn More
SP Bel-Art™ Stirring Bars Spinbar™
Multi-faceted shape provides greater surface area and added turbulence for more vigorous, thorough mixing when compared to the smooth surface of cylindrical bars
Brand:  SP Bel-Art™ F37110-3810
Additional Details : Weight : 2.57980kg
 View more versions of this product
Product Code. 16498485
  • 403.00 NOK / Pack of 10
Estimated Shipment: 16-07-2024
Description
  • With pivot ring for better mixing conditions
  • Alnico magnet encapsulated with PTFE™ brand PTFE coating
  • FDA Grade, USP Class VI
  • Good chemical compatibility
  • Autoclavable
Specifications
Specifications
10 mm
10/Pk.
38 mm
Octagon