Learn More
SP Bel-Art™ Stirring Bars Spinbar™
Multi-faceted shape provides greater surface area and added turbulence for more vigorous, thorough mixing when compared to the smooth surface of cylindrical bars
Brand:  SP Bel-Art™ F37110-5110
Additional Details : Weight : 1.85980kg
 View more versions of this product
Product Code. 16418495
  • 339.00 NOK / Pack of 5
Estimated Shipment: 17-07-2024
Description
  • With pivot ring for better mixing conditions
  • Alnico magnet encapsulated with PTFE™ brand PTFE coating
  • FDA Grade, USP Class VI
  • Good chemical compatibility
  • Autoclavable
Specifications
Specifications
10 mm
5/Pk.
51 mm
Octagon