Learn More
Poulten Graf™ Single Use Luer-Tip Two-Part Syringe
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
10 to 12 mL
2 to 3 mL
20 to 24 mL
5 to 6 mL
50 to 60 mL
missing translation for 'unitSize'
Pack of 100
Pack of 30
Description
- Produced in accordance with DIN 13098
- Available in a variety of sizes, with and without needle
Specifications
Specifications
Autoclavable | Autoclavable |
Certifications/Compliance | CE |
Material | Polypropylene |
Description | Screen printed graduation |
Product Type | Luer-tip 2-part Syringe |
Disposable | Single-use |
Quantity | 100/Pk. |
Volume (Metric) | 5 to 6 mL |
Connection Type | Luer Tip |
Graduated | Yes |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Poulten Graf™ Single Use Luer-Tip Two-Part Syringe >