Learn More
BD Plastipak™ Plastic Concentric Luer-Lock Syringe
Product Code. 10636531
Change view
Click to view available options
Volume (Metric):
20 mL
30 mL
50 mL
Unit Size:
Pack of 120
Pack of 60
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10636531 50 mL Pack of 60
10569215 20 mL Pack of 120
10109922 30 mL Pack of 60
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10636531 Supplier BD Supplier No. 300865
  • ONLINE EXCLUSIVE PRICE
    2205.44 NOK
    708.99 NOK / Pack of 60
    Save 1496.45 NOK
    67% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Enables accurate dosing and visualisation of the syringe contents. BD Medical™ Plastipak™ Plastic Concentric Luer-Lock Syringe are each tested for sterility, pyrogenicity and toxicity.
  • Concentric Luer-Lok
  • Latex free stopper
  • CE marked
  • Polypropylene
  • Sterile
  • Disposable
  • Without needleFor needles, please see our Microlance™ disposable needles range. All products are for research use only, unless otherwise indicated.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE Mark, European Directive 93/42 / EEC (compliance procedure Annex V and VII) Certificates: No. 95 Q 252156 and No. 252231 Q (references 309661 & 309620 ) ; certified by NSAI (No. 0050)
Material Plastic
Color Transparent
Diameter (Metric) Outer 29.4 mm
Latex Free Yes
Length (Metric) 133 mm
Product Type Concentric Luer-lock Syringe
Disposable Single-use
Graduations 1 mL
Quantity Pk. of 60
Volume (Metric) 50 mL
Connection Type Concentric Luer Lock
Graduated Yes