Learn More
Memmert™ Shaking Device for Waterbaths With Support Frame
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
WTB15
WTB24
Unit Size:
Each
Description
- Requires gable cover
- Voltage 230 V, 50/60 Hz
- Order the support basket separately
- Includes support frame, shaking frequency 35 - 160 strokes per minute (horizontal back/forth movement)
Specifications
Specifications
| For Use With (Equipment) | WTB24 |
| Type | Shaking device for waterbaths incl. support frame |
| Speed | 35-160 rpm |
Product Content Correction
Product Title
Memmert™ Shaking Device for Waterbaths With Support Frame >