Learn More
SVL™ Socket/Screw Joint Adapter
Facilitates connection between SVL™ and Quickfit™ glassware.
2905.00 NOK - 7765.00 NOK
Specifications
For Use With (Equipment) | SVL™ and Quickfit™ Glassware |
---|---|
Material | Borosilicate Glass |
Product Type | Socket/screw thread ground joint adapter |
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | |||||
15567740
|
SVL
670-05 |
30 |
7765.00 NOK
Pack of 10 |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
15557740
|
SVL
670-03 |
22 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15547740
|
SVL
670-00 |
15 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Features an SVL™ screwthread and Quickfit™ ground glass socket
- Thread accepts SVL™ screw caps and couplings. Socket fits Quickfit™ cones
- Manufactured from temperature and chemical resistant borosilicate glass
Specifications
SVL™ and Quickfit™ Glassware | |
Borosilicate Glass | |
Socket/screw thread ground joint adapter |