Learn More
SVL™ Socket/Screw Joint Adapter
Facilitates connection between SVL™ and Quickfit™ glassware.
Brand: SVL 670-05
7376.75 NOK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.08000kg
Description
- Features an SVL™ screwthread and Quickfit™ ground glass socket
- Thread accepts SVL™ screw caps and couplings. Socket fits Quickfit™ cones
- Manufactured from temperature and chemical resistant borosilicate glass
Specifications
SVL™ and Quickfit™ Glassware | |
Borosilicate Glass | |
Socket/screw thread ground joint adapter | |
30 |