Learn More
SVL™ Socket/Screw Joint Adapter
Product Code. 15547740
Click to view available options
Standard Taper Joint No.:
15
22
30
Unit Size:
Pack of 10
Product Code. 15547740
Brand: SVL 67000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Facilitates connection between SVL™ and Quickfit™ glassware.
  • Features an SVL™ screwthread and Quickfit™ ground glass socket
  • Thread accepts SVL™ screw caps and couplings. Socket fits Quickfit™ cones
  • Manufactured from temperature and chemical resistant borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) SVL™ and Quickfit™ Glassware
Material Borosilicate Glass
Product Type Socket/screw thread ground joint adapter
Standard Taper Joint No. 15