missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Prostaglandin E2 Parameter Assay Kit

Prostaglandin E2 Parameter Assay Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

5645.00 NOK - 32155.00 NOK

Specifications

Assay Range 39.00 - 2500pg/mL
Assay Sensitivity 41.4pg/mL
Conjugate Biotin
Cross Reactivity Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
Product Type ELISA Kit
View More Specs

Products 2
Product Code Brand Sufficient For Price Quantity & Availability  
Product Code Brand Sufficient For Price Quantity & Availability  
18721854
View Documents
R&D Systems™
KGE004B
1 x 96-well Plate
5645.00 NOK
1 set
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18771934
View Documents
R&D Systems™
SKGE004B
6 x 96-well Plates
32155.00 NOK
1 set
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Prostaglandin E2 Parameter Assay Kit quantifies multi-species Prostaglandin E2/PGE2 in cell culture supernates (51 ul), serum (51 ul), edta plasma (51 ul), heparin plasma (51 ul), urine (51 ul).
Specifications

Specifications

39.00 - 2500pg/mL
Biotin
ELISA Kit
PGE2 Levels in Various Samples
ELISA
Cell Culture Supernates (51uL),Serum (51uL),EDTA Plasma (51uL),Heparin Plasma (51uL),Urine (51uL)
Dinoprostone, PGE2
41.4pg/mL
Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine
Multi-species
1 Kit
Store the unopened product at -20° to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
R&D Systems™ Prostaglandin E2 Parameter Assay Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.