Learn More
Tocris Bioscience™ Procaterol hydrochloride
GREENER_CHOICE
Product Code. 30001408
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 30001408
Brand: Tocris Bioscience™ 1102/10
Potent β2 agonist
Specific and very potent β2 agonist.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 62929-91-3
Molecular Formula C16H23ClN2O3
Specifications
Quantity 10 mg
Formula Weight 326.82
Purity >99%
Molecular Weight (g/mol) 326.821
Chemical Name or Material Procaterol hydrochloride