CAS RN 62929-91-3
CAS RN 62929-91-3
IUPAC Name:
hydrogen 8-hydroxy-5-[(1R,2S)-1-hydroxy-2-[(propan-2-yl)amino]butyl]-1,2-dihydroquinolin-2-one chloride
Synonyms:
procaterol hydrochloride
Molecular Weight (g/mol):
326.82
Molecular Formula:
C16H23ClN2O3
InChi Key:
AEQDBKHAAWUCMT-CVHDTDHSSA-N
SMILES:
[H+].[Cl-].CC[C@H](NC(C)C)[C@H](O)C1=C2C=CC(=O)NC2=C(O)C=C1
1
–
1
of
1
results
1
Tocris Bioscience™ Procaterol hydrochloride
CAS: 62929-91-3 Molecular Formula: C16H23ClN2O3