Learn More
Product Code | Brand | No. of Positions | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Positions | Price | Quantity & Availability | ||||||
11446367
|
Grant Instruments™
PV-32 |
32 |
641.00 NOK 620.04 NOK / Each Save 20.96 NOK 3% Off |
Estimated Shipment: 26-11-2025
Log in to see stock. |
||||||
15251226
|
Grant Instruments™
PV6-10 |
6 |
590.00 NOK 573.24 NOK / Each Save 16.76 NOK 2% Off |
Estimated Shipment: 26-11-2025
Log in to see stock. |
||||||
Description
For use with V-32 Vortex Mixer
Replacement Platform PV-32:
- Fits 3 tube sizes:
- 16 x 1.5mL
- 8 x 0.5mL
- 8 x 0.2mL
Universal 6-socket platform PV-6:
- 6 x 10mL tubes
- 15mm maximum tube dia.
Product Title
Grant Instruments™ Platform For V-32 Vortex Mixer