Learn More
Grant Instruments™ Platform For V-32 Vortex Mixer
Click to view available options
No. of Positions:
6
32
Unit Size:
Each
Description
For use with V-32 Vortex Mixer
Replacement Platform PV-32:
- Fits 3 tube sizes:
- 16 x 1.5mL
- 8 x 0.5mL
- 8 x 0.2mL
Universal 6-socket platform PV-6:
- 6 x 10mL tubes
- 15mm maximum tube dia.
Specifications
Specifications
| For Use With (Equipment) | V-32 Multi Vortex Mixer |
| No. of Positions | 32 |