Learn More
OHAUS™ Module Block
OHAUS™ Module Block features close contact of tubes-to-walls allowing for maximum heat retention and is available with or without a thermometer well.
1614.43 NOK - 3626.62 NOK
Product Code | Brand | Holds | Includes | No. of Places | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Includes | No. of Places | Price | Quantity & Availability | ||||
15722579
|
OHAUS™
30400160 |
1 Block | No Thermometer Well | Solid Block |
1735.00 NOK 1614.43 NOK / Each Save 120.57 NOK 6% Off |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||
15782579
|
OHAUS™
30400166 |
2 Blocks | No Thermometer Well | Solid Block |
2500.00 NOK 2328.06 NOK / Each Save 171.94 NOK 6% Off |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||
15732579
|
OHAUS™
30400161 |
Cuvettes | Thermometer Well | 2 |
2040.00 NOK 1901.05 NOK / Each Save 138.95 NOK 6% Off |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||
15752609
|
OHAUS™
30400193 |
Tubes | Thermometer Well | 9 |
1735.00 NOK 1614.43 NOK / Each Save 120.57 NOK 6% Off |
In Stock
Log in to see stock. |
||||
15782569
|
OHAUS™
30400156 |
Tubes | Thermometer Well | 14 |
1735.00 NOK 1614.43 NOK / Each Save 120.57 NOK 6% Off |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||
15792579
|
OHAUS™
30400167 |
35 mm Tubes | Thermometer Well | 4 |
1735.00 NOK 1614.43 NOK / Each Save 120.57 NOK 6% Off |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||
15772569
|
OHAUS™
30400155 |
25 mm Tubes | Thermometer Well | 6 |
1735.00 NOK 1614.43 NOK / Each Save 120.57 NOK 6% Off |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||
15762569
|
OHAUS™
30400154 |
20 mm Tubes | Thermometer Well | 8 |
1735.00 NOK 1614.43 NOK / Each Save 120.57 NOK 6% Off |
In Stock
Log in to see stock. |
||||
15772609
|
OHAUS™
30400195 |
17 to 18 mm Tubes | Thermometer Well | 12 |
1735.00 NOK 1614.43 NOK / Each Save 120.57 NOK 6% Off |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||
15772579
|
OHAUS™
30400165 |
12 to 13 mm Tubes | No Thermometer Well | 16 |
1735.00 NOK 1614.43 NOK / Each Save 120.57 NOK 6% Off |
In Stock
Log in to see stock. |
||||
15732609
|
OHAUS™
30400191 |
2 mL Tubes | Thermometer Well | 20 |
1735.00 NOK 1614.43 NOK / Each Save 120.57 NOK 6% Off |
In Stock
Log in to see stock. |
||||
15742609
|
OHAUS™
30400192 |
2 mL Corning Tubes | Thermometer Well | 20 |
1735.00 NOK 1614.43 NOK / Each Save 120.57 NOK 6% Off |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||
15742579
|
OHAUS™
30400162 |
11.5/1.5 mL Micta | Thermometer Well | 20 |
1735.00 NOK 1614.43 NOK / Each Save 120.57 NOK 6% Off |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||
15742569
|
OHAUS™
30400152 |
12 to 13 mm Tubes | Thermometer Well | 20 |
1735.00 NOK 1614.43 NOK / Each Save 120.57 NOK 6% Off |
In Stock
Log in to see stock. |
||||
15762609
|
OHAUS™
30400194 |
Microtubes | Thermometer Well | 21 |
1735.00 NOK 1614.43 NOK / Each Save 120.57 NOK 6% Off |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||
15792569
|
OHAUS™
30400157 |
0.5 mL Microtaper | Thermometer Well | 30 |
1735.00 NOK 1614.43 NOK / Each Save 120.57 NOK 6% Off |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||
15752579
|
OHAUS™
30400163 |
250ÎĽL Tubes/6 mm Tubes | Thermometer Well | 30 |
1735.00 NOK 1614.43 NOK / Each Save 120.57 NOK 6% Off |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||
15702579
|
OHAUS™
30400158 |
6 mm Tubes | Thermometer Well | 30 |
1735.00 NOK 1614.43 NOK / Each Save 120.57 NOK 6% Off |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||
15722589
|
OHAUS™
30400170 |
0.2 mL Microtubes | No Thermometer Well | 64 |
1875.00 NOK 1754.82 NOK / Each Save 120.18 NOK 6% Off |
In Stock
Log in to see stock. |
||||
15712589
|
OHAUS™
30400169 |
0.2 mL Microtubes | No Thermometer Well | 80 |
1875.00 NOK 1754.82 NOK / Each Save 120.18 NOK 6% Off |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||
15732589
|
OHAUS™
30400171 |
0.2 mL Microtubes | Thermometer Well | 96 |
3895.00 NOK 3626.62 NOK / Each Save 268.38 NOK 6% Off |
Estimated Shipment: 12-11-2025
Log in to see stock. |
||||
Description
- Block heater accessories include modular blocks to accommodate almost any sample
- Modular blocks feature close contact of tubes-to-walls allowing for maximum heat retention
- Most blocks feature a thermometer well for measuring block temperature
- Low temperature covers help to reduce the air flow for additional temperature stability
Specifications
Anodized Aluminum | |
Block |
Product Title
OHAUS™ Module Block