Learn More
Sampling Systems™ Inserts for use with Multi Thief
Enables the sample volume to be quickly and easily changed by changing the removable sample cells. Sampling Systems™ Insert for use with Multi Thief is ideal for taking multiple bulk samples from free flowing powders and granules.
2334.41 NOK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
15859241
|
Sampling Systems™
1290W-05 |
5 mL |
2570.00 NOK 2334.41 NOK / Each Save 235.59 NOK 9% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15869241
|
Sampling Systems™
1290W-10 |
10 mL |
2570.00 NOK 2334.41 NOK / Each Save 235.59 NOK 9% Off |
Estimated Shipment: 11-12-2025
Log in to see stock. |
||||||
15879241
|
Sampling Systems™
1290W-25 |
25 mL |
2570.00 NOK 2334.41 NOK / Each Save 235.59 NOK 9% Off |
Estimated Shipment: 05-12-2025
Log in to see stock. |
||||||
15889241
|
Sampling Systems™
1290W-50 |
50 mL |
2570.00 NOK 2334.41 NOK / Each Save 235.59 NOK 9% Off |
Estimated Shipment: 11-12-2025
Log in to see stock. |
||||||
15899241
|
Sampling Systems™
1290W-60 |
60 mL |
2570.00 NOK 2334.41 NOK / Each Save 235.59 NOK 9% Off |
Estimated Shipment: 11-12-2025
Log in to see stock. |
||||||
Description
- 316 stainless steel construction
- Variable sample volumes
- Range of sampler lengths
- Suitable for free flowing powders and granules
- Multilevel Powder Sampler with variable Sample Volumes
Specifications
Sampler Insert | |
Stainless Steel (316) |
Product Title
Sampling Systems™ Inserts for use with Multi Thief