Learn More
Sampling Systems™ Inserts for use with Multi Thief
Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
60 mL
Unit Size:
Each
Description
- 316 stainless steel construction
- Variable sample volumes
- Range of sampler lengths
- Suitable for free flowing powders and granules
- Multilevel Powder Sampler with variable Sample Volumes
Specifications
Specifications
| Product Type | Sampler Insert |
| Material | Stainless Steel (316) |
| Capacity (Metric) | 25 mL |