Learn More
Thermo Scientific™ HyPerforma™ Smartainer™ 3.0 Bioprocess Container (BPC), Aegis™ 5-14 film
Shop All Thermo Scientific Products
Description
The Thermo Scientific HyPerforma Smartainer 3.0 BPC provides end-users producing biologics or vaccine therapeutics with a large-volume, sterile liquid storage system for volumes ranging from 200 L to 3000 L—designed for the collection and storage of sterile process liquids such as cell culture media, process water or purification media. Options include temperature-control through integrated water jackets, BPC deploying hoists, as well as other options to optimize performance in your productions processes.
Specifications
Specifications
| Capacity (Metric) | 200 L |
| Product Type | Bioprocess Container (BPC) |
| Insert Type | Horizontally-gusseted |