Learn More
Eppendorf™ Adapters for Combitips Advanced™ Pipet Tips
Shop All Eppendorf Products
Click to view available options
Volume (Metric):
25 mL
50 mL
Color Code:
Light Gray
Red
Quantity:
1 Ea.
7 Pk.
Unit Size:
Each
Pack of 7
Description
Use these adapters with Eppendorf pipet tips. Eppendorf™ Adapters for Combitips Advanced™ Pipet Tips are color-coded and autoclavable.
- Available in 25 and 50mL
Greener Choice Claims
- Environmental benefits include:
- Non-Toxic
- Renewable Energy
- Ozone-Safe or Safe for the Ozone Layer
- Free-of a substance that causes environmental harm
Specifications
Specifications
Volume (Metric) | 25 mL |
Autoclavable | Autoclavable |
Quantity | 1 Ea. |
Color Code | Red |
For Use With (Equipment) | 25 mL Eppendorf Quality Tip |
Promotions
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24158Expires: 2025-10-31
-
Multiple Manufacturers
Focus on Agri-Science Testing
Use promotion code 24136 to get amazing savings.Promo Code: 24136Expires: 2025-10-26
Product Content Correction
Product Title
Eppendorf™ Adapters for Combitips Advanced™ Pipet Tips >