missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ EHNA hydrochloride
GREENER_CHOICE

Product Code. 30001218
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001218 50 mg 50mg
18054892 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001218 Supplier Tocris Bioscience™ Supplier No. 1261/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Adenosine deaminase inhibitor

Selective inhibitor of cGMP-stimulated phosphodiesterase (PDE2) (IC50 = 0.8 - 4 μM). Also a potent inhibitor of adenosine deaminase. Suppresses spontaneous differentiation of human ESCs in feeder-free conditions. Also prevents directed neuronal differentiation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81408-49-3
Molecular Formula C14H24ClN5O

Specifications

Quantity 50 mg
Formula Weight 313.83
Purity >98%
Molecular Weight (g/mol) 313.83
Chemical Name or Material EHNA hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.