accessibility menu, dialog, popup

UserName

H3CH3COHNNNNNH2HCl

CAS RN 81408-49-3

IUPAC Name: 3-(6-amino-9H-purin-9-yl)nonan-2-ol hydrochloride
Synonyms: EHNA hydrochloride
Molecular Weight (g/mol): 313.83
Molecular Formula: C14H24ClN5O
InChi Key: VVDXNJRUNJMYOZ-UHFFFAOYNA-N
SMILES: Cl.CCCCCCC(C(C)O)N1C=NC2=C(N)N=CN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results