Learn More
ABEBA™ Dynamic Anthracite Occupational Clogs Ob ESD
Anthracite dynamic occupational clogs with non-foldable heel strap, replaceable active comfort insole, slip resistant TPU outsole
728.00 NOK - 754.00 NOK
Specifications
Color | Anthracite |
---|---|
Material | Knitted Fabric |
Certifications/Compliance (Europe) | ESD dissipative to EN 61340 Climate class 1, Comply to EN ISO 20347: 2012 OB SRC |
Gender | Unisex |
Ankle Style | Clogs |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17171345
|
ABEBA™
37328/35 |
35 |
728.00 NOK
1 pair |
Estimated Shipment: 26-09-2025
Log in to see stock. |
||||||
17181345
|
ABEBA™
37328/36 |
36 |
728.00 NOK
1 pair |
Estimated Shipment: 26-09-2025
Log in to see stock. |
||||||
17171355
|
ABEBA™
37328/45 |
45 |
728.00 NOK
1 pair |
Estimated Shipment: 26-09-2025
Log in to see stock. |
||||||
17181355
|
ABEBA™
37328/46 |
46 |
728.00 NOK
1 pair |
Estimated Shipment: 26-09-2025
Log in to see stock. |
||||||
17191355
|
ABEBA™
37328/47 |
47 |
728.00 NOK
1 pair |
Estimated Shipment: 26-09-2025
Log in to see stock. |
||||||
17101365
|
ABEBA™
37328/48 |
48 |
728.00 NOK
1 pair |
Estimated Shipment: 26-09-2025
Log in to see stock. |
||||||
17191345
|
ABEBA™
37328/37 |
37 |
728.00 NOK
1 pair |
Estimated Shipment: 26-09-2025
Log in to see stock. |
||||||
17101355
|
ABEBA™
37328/38 |
38 |
728.00 NOK
1 pair |
Estimated Shipment: 26-09-2025
Log in to see stock. |
||||||
17111355
|
ABEBA™
37328/39 |
39 |
728.00 NOK
1 pair |
Estimated Shipment: 26-09-2025
Log in to see stock. |
||||||
17121355
|
ABEBA™
37328/40 |
40 |
728.00 NOK
1 pair |
Estimated Shipment: 26-09-2025
Log in to see stock. |
||||||
17131355
|
ABEBA™
37328/41 |
41 |
754.00 NOK
1 pair |
Estimated Shipment: 26-09-2025
Log in to see stock. |
||||||
17141355
|
ABEBA™
37328/42 |
42 |
728.00 NOK
1 pair |
Estimated Shipment: 26-09-2025
Log in to see stock. |
||||||
17151355
|
ABEBA™
37328/43 |
43 |
728.00 NOK
1 pair |
Estimated Shipment: 26-09-2025
Log in to see stock. |
||||||
17161355
|
ABEBA™
|
44 |
728.00 NOK
1 pair |
Estimated Shipment: 26-09-2025
Log in to see stock. |
||||||
Description
- Breathable, lightweight, comfortable occupational clogs
- Comply to EN ISO 20347: 2012, OB SRC
- ESD dissipative according to DIN EN 61340
- Slip resistant TPU outsole
- Good cushioning
- Energy absorption in heel area
- Water repellent anthracite knitted textile
- Non foldable heel strap, adjustable
- Removable active comfort insole
Specifications
Anthracite | |
ESD dissipative to EN 61340 Climate class 1, Comply to EN ISO 20347: 2012 OB SRC | |
Clogs | |
OB SRC ESD |
Knitted Fabric | |
Unisex | |
Water Resistant |
Product Title
ABEBA™ Dynamic Anthracite Occupational Clogs Ob ESD