Learn More
Tocris Bioscience™ CS 2100
GREENER_CHOICE
Product Code. 30001530
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 30001530
Brand: Tocris Bioscience™ 4543/10
Selective S1P1 agonist
Sphingosine-1-phosphate receptor 1 (S1P1) agonist (EC50 = 4.0 nM). Exhibits 5000-fold selectivity for human S1P1 over S1P3. Displays efficacy in a rat adjuvant-induced arthritis model.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 913827-99-3
Molecular Formula C25H23N3O4S
Specifications
Quantity 10 mg
Formula Weight 461.53
Purity >97%
Molecular Weight (g/mol) 461.536
Chemical Name or Material CS 2100