accessibility menu, dialog, popup

Test

CAS RN 913827-99-3

CAS RN 913827-99-3

IUPAC Name: 1-({4-ethyl-5-[5-(4-phenoxyphenyl)-1,2,4-oxadiazol-3-yl]thiophen-2-yl}methyl)azetidine-3-carboxylic acid
Synonyms: 1-({4-ethyl-5-[5-(4-phenoxyphenyl)-1,2,4-oxadiazol-3-yl]thiophen-2-yl}methyl)azetidine-3-carboxylic acid
Molecular Weight (g/mol): 461.54
Molecular Formula: C25H23N3O4S
InChi Key: DWVJASHDNJMDNH-UHFFFAOYSA-N
SMILES: CCC1=C(SC(CN2CC(C2)C(O)=O)=C1)C1=NOC(=N1)C1=CC=C(OC2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results