Learn More
STEMCELL Technologies Compound E
Notch pathway inhibitor; Inhibits Notch receptor and amyloid precursor protein
Brand:  STEMCELL Technologies 73952
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 17100302
  • 3159.74 NOK / 500µg
Estimated Shipment: 08-05-2024
to see stock.
Description
Description
  • Compound E is a potent, cell-permeable, selective inhibitor that blocks the cleavage of both gamma-secretase and the Notch intracellular domain with similar IC50 values, ranging from 0.24 to 0.37 nM (Beher et al.; Seiffert et al.)
  • Inhibits growth, differentiation, and motility of neuroblastoma cells (Ferrari-Toninelli et al.)
  • Accelerates the differentiation of human embryonic stem cells into primitive neural stem cells (Li et al.)
Specifications
Specifications
209986-17-4
500 μg
≥98%
γ-secretase inhibitor XXI
C27H24F2N4O3