Learn More
Hirschmann™ Class A Kohlrausch Measuring Flasks Product Code.: 11720854
Only undefined left
Add to basket
Hirschmann™ Class A Kohlrausch Measuring Flasks
Product Code. 11720854
Capacity: 100 mL
Click to view available options
:
Capacity: 100 mL
Product Code. 11720854
Brand: Hirschmann™ 2970180
Only undefined left
Add to basket
Only undefined left
Add to basket
For sugar examination pursuant to Kohlrausch.
  • Class A
  • Manufactured from DURAN™ borosilicate glass for resistance to chemicals and thermal shock
  • Certified conformity, with dated batch identification
  • Calibrated for IN
  • Blue graduations
Specifications
Calibration Calibrated for IN
Capacity (Metric) 100 mL
Material Borosilicate Glass
For Use With (Application) Sugar examination
Graduations Blue
Height (Metric) 170 ± 5 mm
Product Type Measuring flask
Tolerance 0.1 mL
Graduated Yes
Class A
Bottom Shape Flat
Product Title
Hirschmann™ Class A Kohlrausch Measuring Flasks >