Learn More
Hirschmann™ Class A Amber Glass White Graduated Measuring Flasks
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
Unit Size:
Pack of 2
Description
- White graduation
- Class A
- With dated batch identification
- Standard ground and PE stopper
- Manufactured from DURAN™ borosilicate glass for high chemical and thermal resistance
- USP certified
Specifications
Specifications
| Closure Material | Polyethylene |
| Color | Amber |
| Capacity (Metric) | 100 mL |
| Material | Borosilicate Glass |
| For Use With (Application) | Liquid handling of light-sensitive products |
| Height (Metric) | 170 ± 5 mm |
| Standard Taper Joint | Ground 12/21 |
| Graduated | Yes |
| Class | A |
| Bottom Shape | Flat |
| Show More |