Learn More
Thermo Scientific™ CaptureSelect™ C-tagXL Pre-packed Column
Shop All Thermo Scientific ProductsDescription
CaptureSelect C-tagXL Affinity Matrix is an improved version of CaptureSelect C-tag Affinity Matrix with a significantly increased binding capacity, offered here in pre-packed format.
Like C-tag Affinity Matrix, C-tagXL Affinity Matrix combines a unique selectivity for a small 4-amino acid peptide tag (E-P-E-A, glutamic acid-proline-glutamic acid-alanine) with the benefits of a robust and high quality affinity matrix. CaptureSelect C-tagXL Affinity Matrix purifies C-terminal tagged proteins with high affinity and selectivity, even in the presence of urea and guanidine HCl, from complex mixtures like cell culture harvests and periplasmatic fractions in a one-step process. Mild elution conditions at neutral pH can be applied using magnesium chloride or propylene glycol, which ensures high activity recoveries of pH-sensitive target proteins. The affinity resin recognizes the E-P-E-A tag sequence when fused either directly to the C-terminus of a protein or through a linker between the C-terminus and the E-P-E-A tag.
The affinity matrix is offered in a pre-packed format as a set of 5 x 1 mL columns, or as one 5 mL column, with connectors that are compatible with standard chromatography equipment.
Specifications
Specifications
Content And Storage | Store in refrigerator (2°C to 8°C). |
Shipping Condition | Room Temperature |
Form | Liquid Suspension |
Product Line | CaptureSelect™ |
Quantity | 1 x 5 mL Column |
Type | Pre-packed Column |
Stationary Phase | Anti-C-tagXL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.