missing translation for 'onlineSavingsMsg'
Learn More

Canertinib dihydrochloride, Thermo Scientific™

Product Code. 17545565 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 17545565

Brand: Thermo Scientific Acros 468930010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 289499-45-2
Molecular Formula C24 H25 Cl F N5 O3 · 2 H Cl

Specifications

Quantity 1 g
Formula Weight 558.87
MDL Number MFCD09954112
Physical Form Powder
Color White to Beige
Purity ≥97.5%
InChI Key JZZFDCXSFTVOJY-UHFFFAOYSA-N
SMILES Cl.Cl.FC1=CC=C(NC2=C3C=C(NC(=O)C=C)C(OCCCN4CCOCC4)=CC3=NC=N2)C=C1Cl
IUPAC Name N-{4-[(3-chloro-4-fluorophenyl)amino]-7-[3-(morpholin-4-yl)propoxy]quinazolin-6-yl}prop-2-enamide dihydrochloride
Molecular Weight (g/mol) 558.86
Chemical Name or Material Canertinib dihydrochloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.