Learn More
Azlon™ Tapered Containers with Snap-On Lid
Constructed of heavy duty, chemically resistant Polypropylene and autoclavable at 121°C Azlon™ Tapered Containers with Snap-On Lid includes a moulded handle for safe and secure carrying. Squat form provides easy location on shelves, bench top, or while in transit. Moulded graduations add quick, easy volume assessment. Caps on 10 & 20 litre carboys have moulded drill guides to aid installation of feed-lines.
1155.00 NOK - 3675.00 NOK
Specifications
For Use With (Application) | Storage of Samples, Powders, Animal Feeds |
---|---|
Temperature Range (Metric) | –20 to 135°C |
Closure Included | Yes |
Autoclavable | Autoclavable |
Mouth | Wide |
Product Code | Brand | Capacity (Metric) | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Includes | Price | Quantity & Availability | |||||
12076892
|
Azlon
JWH0125P |
125 mL | Snap-On Lid |
3675.00 NOK
Pack of 250 |
Estimated Shipment: 10-10-2025
Log in to see stock. |
|||||
12086892
|
Azlon
JWH0250P |
250 mL | Tight Fitting LDPE Lid |
2450.00 NOK
Pack of 250 |
Estimated Shipment: 24-10-2025
Log in to see stock. |
|||||
12096892
|
Azlon
JWH0500P |
400 mL | Tight Fitting LDPE Lid |
1390.00 NOK
Pack of 100 |
Estimated Shipment: 17-10-2025
Log in to see stock. |
|||||
12006902
|
Azlon
JWH1000P |
1000 mL | Tight Fitting LDPE Lid |
2295.00 NOK
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||||
12016902
|
Azlon
JWH2000P |
2000 mL | Tight Fitting LDPE Lid |
2625.00 NOK
Pack of 50 |
Estimated Shipment: 25-09-2025
Log in to see stock. |
|||||
12585087
|
Azlon
JWH2500P |
2500 mL | Tight Fitting LDPE Lid |
1155.00 NOK
Pack of 25 |
Estimated Shipment: 24-10-2025
Log in to see stock. |
|||||
12036902
|
Azlon
JWH5000P |
5000 mL | Tight Fitting LDPE Lid |
1735.00 NOK
Pack of 25 |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
Description
- Useful lightweight tapered containers with tight-fitting LDPE lids
- Good chemical resistance
- Ideal for storage of samples, powders, animal feeds, and more
- Suitable for food contact applications in accordance with USA regulations
Specifications
Storage of Samples, Powders, Animal Feeds | |
Yes | |
Wide | |
Storage Container | |
Snap Lid |
–20 to 135°C | |
Autoclavable | |
Round | |
LDPE |