Learn More
Azlon™ Tapered Containers with Snap-On Lid
Click to view available options
Capacity (Metric):
125 mL
250 mL
400 mL
1000 mL
2000 mL
2500 mL
5000 mL
Includes:
Snap-On Lid
Tight Fitting LDPE Lid
Unit Size:
Pack of 100
Pack of 25
Pack of 250
Pack of 50
Description
- Useful lightweight tapered containers with tight-fitting LDPE lids
- Good chemical resistance
- Ideal for storage of samples, powders, animal feeds, and more
- Suitable for food contact applications in accordance with USA regulations
Specifications
Specifications
| Capacity (Metric) | 400 mL |
| For Use With (Application) | Storage of Samples, Powders, Animal Feeds |
| Includes | Tight Fitting LDPE Lid |
| Material | HDPE |
| Quantity | 100/PK |
| Temperature Range (Metric) | –20 to 135°C |
| Weight (Metric) | 3.13 kg |
| Closure Included | Yes |
| Mouth | Wide |
| Shape | Round |
| Show More |